missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ MAS™ Bilirubin Controls
Monitor the performance of total, conjugated, unconjugated and neonatal Bilirubin with tri-level, assayed MAS Bilirubin Controls.
£272.00
Specifications
Content And Storage | Two year shelf life from date of manufacture at 2° to 8°C; 14 days open vial at 2° to 8°C when stored in the dark |
---|---|
Description | Bilirubin Control, Assayed |
Detectable Analytes | Bilirubin Conjugated, Bilirubin Total (TBIL), Bilirubin Direct (DBIL), Bilirubin Unconjugated, Bilirubin Neonatal |
Sample Type | Serum |
Type | Control |
Description
Monitor the performance of total, conjugated, unconjugated and neonatal Bilirubin with tri-level, assayed Thermo Scientific™ MAS™ Bilirubin Controls. These controls are also available in a convenient Vista™ vial format.
- Liquid, ready-to-use; no additional preparation steps required
- Bovine serum matrix
- Assayed values available for most major analyzers
- Elevated bilirubin in Level 3 for neonatal applications
Specifications
Two year shelf life from date of manufacture at 2° to 8°C; 14 days open vial at 2° to 8°C when stored in the dark | |
Bilirubin Conjugated, Bilirubin Total (TBIL), Bilirubin Direct (DBIL), Bilirubin Unconjugated, Bilirubin Neonatal | |
Control | |
5 mL |
Bilirubin Control, Assayed | |
Serum | |
6 Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ MAS™ Bilirubin Controls