Learn More
Bolle Safety Masterall Safety Goggles
Click to view available options
Double lens
Description
- Equalizer technology filters and regulates moisture levels in goggles
- Permanent scratch-resistant platinum coating
- Sealed PVC frame and neoprene strap
- For use in offshore and petrochemical environments
Specifications
Lens Coating | Scratch-resistant |
Description | Double lens |
Product Type | Safety goggles |
For Use With (Application) | Offshore and petrochemical environments |
Frame Type | Sealed PVC |