Learn More
BarbLock Assemblies
Brand: Masterflex
Code : NEW
Additional Details : Weight : 0.01000kg
Specifications
BarbLock Assemblies | |
Fits 1/8 in. x 1/4 in. Tubing | |
Easy to install, three-piece locking mechanism ensures the tubing and hose barb won't disengage | |
1 Pack |