missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ BioPharm Plus™ I/P™ High-Performance Precision Pump Tubing, Platinum-Cured Silicone

Ensure your tubing is clean, free from defects and performs to exact specifications. Masterflex™ BioPharm Plus™ I/P™ High Performance Precision Pump Tubing is ideal for applications involving pressure, suction lift, viscous fluids or long tubing life.

Brand:  Masterflex™ 96441-89

 View more versions of this product

Product Code. 11897523

  • £1255.00 / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

Tubing is manufactured to tight tolerances—ensures accurate flow rates and a long tubing life. Firm tubing has longer life, better pressure generation, a stronger vacuum/suction lift and lower gas permeability than softer tubings.
  • Good chemical resistance
  • Resists corrosives, solvents and oils at elevated temperatures
  • Meets USP Class VI criteria, and is FDA compliant
  • Optically inspected and factory tested
  • Nontoxic and nonhemolytic with excellent biocompatibility
  • Long pumping life; pump at continuous pressures up to 40 psi
  • Low extractables and low gas permeability
Specifications

Specifications

Pump Tubing
15.9 mm
300 cm
Organic Solvents
0.03 to 19L/min.
8.2 m
610 mm
Autoclavable
Platinum-cured Silicone
-59°C
89
5/8in.
20 psi
232°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers