missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ HDPE Barbed Fitting

Ensure a tight seal every time with Masterflex fittings. Designed to tight tolerances, these fittings match perfectly to your tubing to ensure smooth flow and a proper fit.

Brand:  Masterflex™ 30615-05

 View more versions of this product

Product Code. 12334598

  • £109.00 / Pack of 10
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Requires no tools for quick and easy installation
  • Temperature range from −53° to +65°C
  • Very good chemical resistance
  • Sterilize by ethylene oxide only
  • A variety of shapes and sizes available
  • 10/pack
Specifications

Specifications

Barbed Fitting
0.125 in. Tubing I.D.
10 Pack
ETO sterilization
HDPE
-53°C
Straight
65°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers