missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Masterflex™ I/P™ Digital Process Drive Systems

Product Code. 16418675
Click to view available options
Flow Rate:
0.001 to 8 LPM
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16418675

Brand: Masterflex™ 7796428

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Maintenance-free drives built for precision dispensing.

  • Flow rate 0.001 to 8 LPM with included I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in mL (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
  • Rugged housing is NEMA 4X (IP66) rated for washdown
  • Choose models with open head sensor technology for enhanced user safety

Specifications

Display Type LCD
Enclosure Rating IP66
Flow Rate 0.001 to 8 LPM
Motor Speed 0.1 to 650 rpm
Includes I/P drive 77420-18, Easy-Load pump head 77602-30, 10-ft of C-Flex ULTRA™ tubing, and 6 ft. (1.8 m) line cord with IEC 320/CEE22 socket
Material Stainless Steel
Electrical Requirements 90 to 260 VAC
Amperage 4.5 A, 2.5 A
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.