Learn More
Masterflex™ I/P™ Digital Modular Drive with Benchtop Controller and Remote Input/Output
Place motor and controller where convenient with modular design
Brand:  Masterflex™ 07591-20
Product Code. 13447099
  • ONLINE EXCLUSIVE PRICE
    ÂŁ4455.00
    ÂŁ3860.00 / Each
    Save ÂŁ595.00
    13% Off
Only null left
Estimated Shipment: 05-02-2026
Add to basket
Description
Description
  • Flow rate 0.04 to 19 LPM with I/P™ tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with LED display shows pump rpm— confirm speed setting at a glance
  • Remote control available using analog communication—voltage, current and contact closure
  • Powerful motor lets you stack two pump heads for increased flow rate while offering ±0.25% speed control accuracy
Specifications
Specifications
115/230 VAC 149 w
6 ft. (1.8-m) line cord with IEC 320/CEE22 socket
39.5 cm
Yes
149 w
19,000 mL/min.
15.2 cm
14.1 cm
0.25%
6 to 650 rpm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Masterflex™ I/P™ Digital Modular Drive with Benchtop Controller and Remote Input/Output > Flow Rate: 19000mL/min.