Learn More
Sartorius™ Masterflex™ Economy Drive Peristaltic Pump
Optimally suited to run Vivaflow™ laboratory crossflow cassette units. Sartorius™ Masterflex™ Economy Drive Peristaltic Pump is durable and cost-effective.
Brand: Sartorius™ VFP001
1195.60 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- Suitable for Vivaflow™ laboratory crossflow cassette units
- Durable and affordably priced
- Variable speed
Specifications
Variable | |
4.1 kg | |
230 V | |
37 w |
Product Content Correction
Product Title
Sartorius™ Masterflex™ Economy Drive Peristaltic Pump > Dimensions (L x W x H): 220 x 180 x 135mm
Spot an opportunity for improvement?Share a Content Correction