Learn More
Masterflex™ L/S™ High-Performance Precision Pump Tubing, Platinum-Cured Silicone
Excellent biocompatibility that meets multiple classifications
£180.00 - £231.00
Product Code | Brand | Tubing Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | ||||||
11855181
|
Masterflex™
96410-15 |
15 |
£180.00
7.60m |
In Stock
Log in to see stock. ![]() |
||||||
11720989
|
Masterflex™
96410-35 |
35 |
£203.00
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
10709874
|
Masterflex™
96410-36 |
36 |
£231.00
7.60m |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
Description
Ensure optimal performance from your Masterflex™ pump with this silicone pump tubing. Masterflex™ L/S™ Peroxide-Cured Silicone High-Performance Precision Pump Tubing provides accurate, repeatable flow. Ideal for pharmaceutical and biotechnology, peristaltic pumping and food applications.
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory Approvals to match your unique application
- Always in stock — most formulations and sizes available to ship same day
- Economical tubing with a longer tubing life
Specifications
Autoclavable | |
Platinum-cured Silicone |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Masterflex™ L/S™ High-Performance Precision Pump Tubing, Platinum-Cured Silicone