missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Masterflex™ Polypropylene Masterflex Barbed Fitting
Click to view available options
For Use With:
3/8 in. x 1/4 in. ID Tubing
Unit Size:
Pack of 10
Specifications
Specifications
| Material | Polypropylene |
| For Use With | 3/8 in. x 1/4 in. ID Tubing |
| Packaging Quantity | 10 Pack |
| Color | Red |
| Product Type | Barbed Fitting |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction