Learn More
Masterflex™ L/S™ Easy-Load™ II Pump Head Mounting Hardware
Mount up to four pump heads on a single drive. Masterflex™ Stainless Steel Mounting Hardware is designed for use with L/S™ Easy-Load™ II Pump Heads.
Brand:  Masterflex™ 77200-01
Code : XX
Additional Details : Weight : 0.01600kg
 View more versions of this product
Product Code. 11721019
  • £60.10 / Each
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
Description
Description
Mount up to four pump heads on a single drive. Masterflex™ Stainless Steel Mounting Hardware is designed for use with L/S™ Easy-Load™ II Pump Heads.
  • For use with L/S Easy-Load II Pump Heads
  • See individual drive specifications for maximum number of heads to be mounted
  • Mounting hardware is stainless steel
Specifications
Specifications
Mounting one L/S Easy-Load II pump head
2 Pack
Stainless Steel
L/S Easy-Load II pump heads
Mounting Hardware