missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ Easy-Load™ II Pump Head with 2-Channels for Precision Pump Tubing

No need to stack heads—get two channels in one head

Brand:  Masterflex™ 77202-60

Product Code. 11883642

  • £731.00 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

No need to stack heads—get two channels in one head

  • Two-channel head lets you pump two synchronous flow channels (proper retention requires the use of the same size tubing in each channel)
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation.
  • Improved occlusion bed geometry lengthens tubing life.
  • Polyphenylene sulfide (PPS) housing with either stainless steel (SS) or cold-rolled steel (CRS) rotor — choose CRS for economy or SS for greater durability and chemical resistance.
  • Each pump head for precision pump tubing accepts tubing sizes L/S13, L/S 14, L/S 16, and L/S 25. Achieve flow rates of 0.06 to 1000mL/min (1 to 600rpm).
Specifications

Specifications

0.06 to 1000 mL/min.
2
Fixed
12.1 cm
Stainless Steel
Pump Head
1 to 600 rpm
77202-60
4
7 cm
12.7 cm
L/S 13, L/S 14, L/S 16, L/S 25
Stainless Steel
Polyphenylene Sulfide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.