Learn More
Masterflex™ MasterflexLive™ L/S™ Cloud-Enabled Digital Process Drive
Monitor and control your pump from anywhere and at any time when using your MasterflexLive-equipped pump drive! Using cloud-based technology, you can monitor and control your pump using your smart phone as an interface, freeing you from being physically tethered to your application.
Description
Description
  • MasterflexLive is a secure, cloud-enabled platform for control and monitoring of Masterflex pumps
  • Get real-time control of all pump parameters – including speed, flow rate, dispense volume, and more
  • Push notifications alert you to operating conditions and error messages
  • Monitor and adjust critical processes running 24/7 without the need for onsite personnel
  • Internet connection via standard Ethernet (RJ45) jack on rear of drive
  • Flow rate 0.006 to 3400 mL/min with L/S tubing. Flow rate depends on drive rpm and tubing size
  • 316 stainless steel housing
  • IP66 and NEMA 4X rated sealed housing and keypad are protected from dust and water in the washdown environment—simply hose down to clean
  • Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense. View pump performance continually
  • Open-head sensor stops the drive when the pump head is opened (when used with sensor-equipped pump head)
  • Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Masterflex™ MasterflexLive™ L/S™ Cloud-Enabled Digital Process Drive