missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAT2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55309-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to MATA2.
The protein encoded by this gene catalyzes the production of S-adenosylmethionine (AdoMet) from methionine and ATP. AdoMet is the key methyl donor in cellular processes.Specifications
MAT2A | |
Polyclonal | |
Unconjugated | |
MAT2A | |
AdoMet synthase 2, AMS2, MAT 2, MAT II, MAT2A, MATA2, MATII, SAMS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
232087, 4144 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P31153, Q3THS6 | |
MAT2A | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction