missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Matrigel™ Growth Factor Reduced (GFR) Basement Membrane Matrix, LDEV-Free
Suited for applications requiring highly defined basement membrane preparation.
£2013.32 - £3921.57
Specifications
Content And Storage | Store at -20°C. Avoid multiple freeze-thaws. Do not store in frost-free freezer. Keep frozen. |
---|---|
Without Additives | No LDEV |
For Use With (Application) | Cell Growth and Differentiation, Metabolism/Toxicology Studies, Invasion Assays, In Vitro and In Vivo Angiogenesis Assays, In Vivo Angiogenesis Studies and Augmentation of Tumors in immunosuppressed Mice |
Source | Engelbreth-Holm-Swarm (EHS) Mouse Tumor |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15505739
|
Corning™
356252 |
50 mL (10 mL x 5) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15515739
|
Corning™
356253 |
100 mL (10 mL x 10) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
Store at -20°C. Avoid multiple freeze-thaws. Do not store in frost-free freezer. Keep frozen. | |
No LDEV | |
Cell Growth and Differentiation, Metabolism/Toxicology Studies, Invasion Assays, In Vitro and In Vivo Angiogenesis Assays, In Vivo Angiogenesis Studies and Augmentation of Tumors in immunosuppressed Mice | |
Engelbreth-Holm-Swarm (EHS) Mouse Tumor |