missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Matrigel™ hESC-Qualified Matrix, LDEV-Free
Click to view available options
Quantity:
25 mL (5 mL x 5)
50 mL (5 mL x 10)
Unit Size:
Each
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
Specifications
| Product Type | hESC-qualified Matrix |
| Without Additives | No LDEV |
| For Use With (Application) | Cell Growth and Differentiation, Metabolism/Toxicology Studies, Invasion Assays, In Vitro and In Vivo Angiogenesis Assays, In Vivo Angiogenesis Studies and Augmentation of Tumors in immunosuppressed Mice |
| Quantity | 25 mL (5 mL x 5) |
| Source | Engelbreth-Holm-Swarm Mouse Tumor |
| Content And Storage | Store at -20°C. Avoid multiple freeze-thaws. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction