Learn More
Corning™ Matrigel™ Matrix-3D Plate, 384-well
GREENER_CHOICE
Corning™ Matrigel™ Matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma. It has a high-throughput format that can be used for drug discovery with 3D cell cultures such as organoids, spheroids or tumoroids.
Brand:  Corning™ 356258
 View more versions of this product
Product Code. 17243494
  • £282.00 / Each
Estimated Shipment: 14-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A high-throughput pre-coated format that contains Matrigel™ and can be used for drug discovery applications with 3D cell cultures such as organoids, spheroids or tumoroids
  • Extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors
  • Each lot is verified for intraplate consistency
  • Tested and found negative for the presence of bacteria and fungi
  • Easy and convenient as Matrigel™ is pre-dispensed into each well
  • Formats provide intra-plate consistency
  • Allows for sandwich/overlay and embedded 3D application workflows
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Barcode
With Lid
Not Autoclavable
Single-use
384
Flat Bottom (F-well)
3D cell culture
1
RUO
No
No
Store at -20°C
White, Clear
Polystyrene
Matrigel Matrix
No
Guidelines for Use
1
No
White with clear bottom
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Matrigel™ Matrix-3D Plate, 384-well > Color: White, Clear; Quantity: 1/Pk, 1/Cs