Learn More
Thermo Scientific™ Matrix™ 2D Barcoded Open-Top Storage Tubes
GREENER_CHOICE
Store and track sample material in these 2D barcoded clear polypropylene open-top storage tubes.
Brand:  Thermo Scientific™ 3750
428.43 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16247831
Description
Show More
Specifications
Specifications
500 μL
Polypropylene
V-bottom
Matrix™
16907 fl oz.
V-bottom
Bulk, 960 Tubes
2D BarCoded
Non-sterile
PCR Ready
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Matrix™ 2D Barcoded Open-Top Storage Tubes >