missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ DeepWell™ Storage Blocks
GREENER_CHOICE

Product Code. 10101303
Click to view available options
Sterility:
Non-sterile
Sterile
Well Shape:
Round
Square, Conical Bottom (V-well)
Volume (Metric) Well:
240 μL
1200 μL
2200 μL
Unit Size:
Pack of 40
Pack of 80
This item is not returnable. View return policy

Product Code. 10101303

Brand: Thermo Scientific™ 421211

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these DeepWell™ storage blocks as mother plates when creating compound libraries from automated liquid handling instruments.

When creating compound libraries using automated liquid handling instruments, use Thermo Scientific™ Matrix™ DeepWell™ Storage Blocks as a mother plate. These strong blocks are constructed of polypropylene and are chemically resistant, meeting all standard microplate specifications. The unique chimney well design of the 1mL blcok reduces the risk of cross-contamination between wells during pipetting, while the 2mL block has cut-away sides for easy handling and placement on robotic systems.

Features:

  • Alphanumeric notations simplify the identification of wells
  • Industry-standard footprint ensures compatibility with sealers, stackers and liquid handlers
TRUSTED_SUSTAINABILITY

Specifications

Barcode Custom Barcoding Available
Certifications/Compliance Certified RNase and DNase Free. ANSI Compliant.
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 40 Pk., 40 Cases
Binding Type Low binding PP
Product Line Nunc
Color Natural
Product Type Polypropylene Storage & Sample Prep Plate
For Use With (Equipment) ALPS Heat Sealers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
Volume (Metric) Well 1200 μL
No. of Wells 96
Volume (Metric) Working 1000 μL
Sterility Sterile
Surface Treatment Non-treated
Well Shape Round
No. per Case 40
No. per Pack 40
Regulatory Status For Research Use Only. Not for use in diagnostic procedures.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.