Learn More
IKA™ MATRIX Delta Plus
MATRIX Delta Plus heats and cools reliably with variable inserts. Whether blood samples, pharmaceutical agents, DNA/RNA samples or ELISA assays - reaction vials and plates with even the smallest volumes are reliably tempered in all laboratory applications.
Brand: IKA™ 0030000717
1843.00 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 8.80000kg
Description
- Large and clear display with a convenient menu structure allows for an intuitive operation
- Reduced waiting time through fast heating and cooling rates of up tp 5.5°C/min (heating) or 2°C/min (cooling, at set-point temperature between ambient temperature and 30°C below ambient temperature)
- Firm quick-release fastener for quick and easy changing of the inserts/attachments as well as an automatic attachment detection
- Automatic performance of heating and cooling tasks through programmable rampsAluminum die cast housing ensures secure positioning, excellent stability and everyday robustness
The device is ideally suited for numerous applications in laboratories, such as:
- Tempering of samples
- Synthesis of cDNA
- Enzyme-catalysed linking of RNA or DNA (ligation)
- Gentle thawing of sample material
- Structural change of DNA, RNA and proteins (denaturation)
- ELISA assays
Specifications
0.3 kg | |
EU Plug | |
1°C | |
50/60 Hz | |
351 x 216 x 127 mm | |
TFT | |
80% | |
Exchangable attachments |
-10°C to +110°C | |
0.5°C | |
0.5°C | |
Aluminum die-cast housing | |
9.5 kg | |
0030001253 MATRIX Cover, 0030001244 MATRIX ThermoCover, 0030000853 MATRIX PCR 96 Attachment, 0030000857 MATRIX PCR 384 Attachment, 0030000840 MATRIX 0.5 mL Attachment, 0030000829 MATRIX 1.5 mL Attachment, 0030000834 MATRIX 2.0 mL Attachment, 0030000859 MATRIX 5 mL Attachment, 0030000844 MATRIX 12 mL Attachment, 0030000821 MATRIX 15 mL Attachment, 0030000863 MATRIX 50 mL Attachment, 0030000848 MATRIX Cryo Attachment | |
Power cable; USB cable | |
0 to 6000 min. |