missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ MATRIX Orbital
IKA MATRIX Orbital mixes and is suitable for variable inserts. Whether its blood samples, pharmaceutical agents, DNA/RNA samples or ELISA assays – samples with even the smallest volumes mix reliably and completely in all laboratory applications.
Brand: IKA™ 0030000627
1271.30 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Alert:
MATRIX, Orbital, T10, AC10
Description
- Large and clear display with a convenient menu structure allows for intuitive operation
- Speeds of up to 3,000 rpm and shaker diameter of 3 mm ensure the liquid mixes quickly and effectively
- Two different speed modes for optimal mixing
- Speed adjustment according to insert, attachment and filling volume
- Aluminum die cast housing ensures secure positioning, excellent stability and everyday robustness
- No cross-contamination and an optimal mixing result
Specifications
3 mm | |
15 rpm Steps | |
EU Plug | |
Aluminum die-cast housing | |
TFT | |
Power cable; USB cable | |
0 to 6000 min. |
0.3 kg | |
0 to 3000 rpm | |
50/60 Hz | |
7.2 kg | |
80% | |
Fixed attachment |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction