missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ MATRIX Orbital Delta FP Thermoshaker
The MATRIX Orbital Delta FP Thermoshaker mixes, heats, and has a universal holder for microtiter plates. Whether its blood samples, pharmaceutical agents, DNA/RNA samples, or ELISA assays, samples with even the smallest volumes mix reliably and completely in all laboratory applications.
Brand: IKA™ 0020025679
This item is not returnable.
View return policy
Description
- Large and clear display with a convenient menu structure allows for intuitive operation
- Quick lock system for fast and easy change of inserts and attachments as well as automatic attachment detection
- Speeds of up to 2000 rpm and shaker diameter of 3 mm ensure the liquid mixes quickly and effectively
- Two different speed modes for gentle heating of samples
- Programmable ramps allow for automatic execution of mixing and heating tasks
- Aluminum die cast housing ensures secure positioning, excellent stability, and general robustness
Specifications
3 mm | |
±30 rpm | |
100/240 V | |
240 W | |
CE | |
±0.5°C | |
9.2 kg | |
MATRIX Orbital Delta FP | |
80% |
0.3 kg | |
0 to 2000 rpm | |
100°C (Max.) | |
Orbital Shaker with Heating | |
UK Plug | |
351 x 216 x 147 mm | |
Touch Key Interface | |
TFT LCD | |
Fixed Universal Holder for Microtiter Plates |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction