missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ 384 Format Pipette Tips

Choose from a comprehensive selection of pipette tips.

£585.36 - £852.12

Specifications

Filtered Non-filtered
Format Rack
For Use With (Equipment) Thermo Scientific Matrix Pipette
Product Line Matrix
View More Specs

Products 4
Product Code Brand Volume (Metric) Sterility Price Quantity & Availability  
Product Code Brand Volume (Metric) Sterility Price Quantity & Availability  
10640784
View Documents
Thermo Scientific™
7421
12.5 μL Non-sterile N/A N/A N/A
10714444
View Documents
Thermo Scientific™
7422
12.5 μL Sterile N/A N/A N/A
10756544
View Documents
Thermo Scientific™
7431
30 μL Non-sterile N/A N/A N/A
10456133
View Documents
Thermo Scientific™
7432
30 μL Sterile N/A N/A N/A
Description

Description

Choose from a comprehensive selection of pipette tips. Thermo Scientific™ Matrix 384 Format Pipette Tips are supplied in innovative new Thermo Scientific™ Flo-Thru™ racks. Matrix pipetters and pipet tips are designed together to ensure that a perfect balance exists between security of the seal and ease of application/ejection. Manufactured under tight QC specifications, these standard tips rival special low-retention tips currently on the market after pipetting fluids such as dH2O, 50% Glycerol, Hybridized Buffer and 0.1% SDS.

Benefits:

  • Sealing rings on tips ensure confident, leak-free seal without banging tips on pipetter.
  • Because the sealing rings compress, these tips also offer a secure seal on most other pipetters on the market.
  • Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives.
  • Stabilization rings in racks hold tips steady when seating on pipetter.
  • Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips.
  • Finger notch on racks allows easy removal/insertion of refill trays.

Innovative Racks and Packaging:

  • Rigid rack design ensures solid, even tip seating; rack will not flex or bend when trying to apply tips.
  • Unique Thermo Scientific™ Flo-Thru™ rack individually supports tips to prevent them from wobbling when trying to seat them on the pipetter.
  • Hinge prevents sterile lid from coming in contact with lab bench.
  • Venting along the edge or around each tip improves heat flow during autoclaving; reduces condensation that can form on the tips.
  • Dual-position lid can be propped open to prevent vacuum deformation during autoclaving.
  • Sterile racks are sold in individual bags with quick-tear openings; each rack remains sterile until ready for use.
  • Each rack is labeled with product details including re-ordering information, description, volume range, and lot number.
  • Racks are constructed of clean recycled materials and are made as small as possible to reduce waste.
  • Racks are marked with appropriate symbols to support recycling initiativ
Specifications

Specifications

Non-filtered
Rack
Thermo Scientific Matrix Pipette
Matrix
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Matrix™ 384 Format Pipette Tips

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.