missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ Pipette Tips

Product Code. 10090754
Click to view available options
Volume (Metric):
10 μL
20 μL
125 μL
250 μL
300 μL
1250 μL
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 3840
Pack of 500
Pack of 960
This item is not returnable. View return policy

Product Code. 10090754

Brand: Thermo Scientific™ 7610

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Choose from a comprehensive selection of these pipette tips designed together with pipetters to ensure a perfect balance between security of the seal and ease of application/ejection.

Choose from a comprehensive selection of pipette tips. Thermo Scientific™ Matrix Pipette Tips are supplied in innovative new Thermo Scientific™ Flo-Thru™ racks. Matrix pipetters and pipet tips are designed together to ensure that a perfect balance exists between security of the seal and ease of application/ejection. Manufactured under tight QC specifications, these standard tips rival special low-retention tips currently on the market after pipetting fluids such as dH2O, 50% Glycerol, Hybridized Buffer and 0.1% SDS.

Benefits:

  • Sealing rings on tips ensure confident, leak-free seal without banging tips on pipetter.
  • Because the sealing rings compress, these tips also offer a secure seal on most other pipetters on the market.
  • Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives.
  • Stabilization rings in racks hold tips steady when seating on pipetter.
  • Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips.
  • Finger notch on racks allows easy removal/insertion of refill trays.

Innovative Racks and Packaging:

  • Rigid rack design ensures solid, even tip seating; rack will not flex or bend when trying to apply tips.
  • Unique Thermo Scientific™ Flo-Thru™ rack individually supports tips to prevent them from wobbling when trying to seat them on the pipetter.
  • Hinge prevents sterile lid from coming in contact with lab bench.
  • Venting along the edge or around each tip improves heat flow during autoclaving; reduces condensation that can form on the tips.
  • Dual-position lid can be propped open to prevent vacuum deformation during autoclaving.
  • Sterile racks are sold in individual bags with quick-tear openings; each rack remains sterile until ready for use.
  • Each rack is labeled with product details including re-ordering information, description, volume range, and lot number.
  • Racks are constructed of clean recycled materials and are made as small as possible to reduce waste.
  • Racks are marked with appropriate symbols to support recycling initiatives.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Autoclavable Autoclavable
Format Bulk
For Use With (Equipment) Thermo Scientific Matrix
Product Line Matrix
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.