Learn More
Thermo Scientific™ Matrix™ Reagent Reservoirs
Accommodate a variety of applications with these uniquely designed, innovative reagent reservoirs.
£87.70 - £218.00
Product Code | Brand | Capacity (Metric) | Material | Packaging | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Material | Packaging | Type | Price | Quantity & Availability | |||
10795393
|
Thermo Scientific™
8075 |
75 mL | Polypropylene | Reagent Reservoir |
£168.00
Pack of 10 |
In Stock
Log in to see stock. |
||||
10495973
|
Thermo Scientific™
8076 |
75 mL | Polypropylene | 10 pcs /bag | Reservoir Lid |
£87.70
Pack of 10 |
In Stock
Log in to see stock. |
|||
10141873
|
Thermo Scientific™
8085 |
100 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir |
£218.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
10375023
|
Thermo Scientific™
8086 |
100 mL | Polystyrene | 50 Individually wrapped | Reagent Reservoir |
£211.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
10756354
|
Thermo Scientific™
8093-11 |
25 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir |
£218.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
10080263
|
Thermo Scientific™
8094 |
25 mL | Polystyrene | 50 Individually wrapped | Reagent Reservoir |
£206.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
10225743
|
Thermo Scientific™
8095 |
25 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir with Divider |
£218.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
10435963
|
Thermo Scientific™
8096-11 |
25 mL | Polystyrene | 50 Individually wrapped | Reagent Reservoir with Divider |
£206.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
Description
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
- Trough within a trough maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
- Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
- Graduations on inside wall enable quick measurement of remaining liquid
- Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open