Learn More
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)
Achieve ultimate performance in automated pipetting with these tips--superior quality with the flexibility to choose tips for any application.
Brand:  Thermo Scientific™ 5572
189.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12917207
  • £259.00 / Pack of 1920
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.
Features:
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
  • Face seal design ensures consistent tip-to-tip height for precise low volume dispensing
  • Highly polished molds and cores ensure low liquid retention.
  • Small internal tip orifice provides better small volume droplet formation, resulting in optimal accuracy and precision.
  • Sterile, sterile filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
Includes: 20 magazines per case
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x2, Matrix™ PlateMate™ 2x3, PlateMate™ Plus Systems and Versette.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 μL
Sterile
Not Autoclavable
Free of RNase, DNase, DNA, ATP and endotoxin
Automation
Store at room temperature.
Single-use
D.A.R.T.S
Non-filtered
Natural
ISO 9001 and ISO 13485
Rack
20 x 96 tips
Room Temperature
Thermo Scientific Matrix PlateMate, Thermo Scientific Matrix Hydra DT