missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ TallTip™ Extended Length Pipette Tips

Product Code. p-4525536
Click to view available options
Volume (Metric):
30 μL
250 μL
300 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Extended Length
Universal
Format:
Bagged
Bulk
Rack, 10 x 96
Unit Size:
Pack of 1000
Pack of 500
Pack of 960
This item is not returnable. View return policy

Product Code. 10690214

Brand: Thermo Scientific™ 7080

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Maximize reach with these extended length pipette tips that allow access to the bottom of test tubes, bottles and flasks.

Maximize reach with Thermo Scientific™ Matrix™ TallTip™ Pipette Tips, which allow access to the bottom of test tubes, reagent bottles, flasks and other vessels without touching the barrel of the pipetter against the side of the tube. Nonbeveled, narrow tip ends provide cleaner touch-off of sidewalls for greater precision.

Benefits:
  • Prevent cross-contamination from vessel walls
  • Allow the use of large volume tips in small spaces resulting in less liquid displacement
  • 102 mm maximum length offers safe access to 100 mm test tubes
  • Thermo Scientific™ Flo-Thru™ rack reduces tip wobble and improves airflow while autoclaving
  • Universal fit works with most major brands of pipetters

Matrix Pipet Tip Features

  • Sealing rings on tips ensure a confident seal without banging tips on pipetter
  • Highly polished molds and rigorous maintenance procedures ensure low liquid retention tips without special treatments or additives
  • Stabilization rings in racks hold tips steady when seated on a pipetter
  • Rigid rack design ensures solid, even tip seating
  • Venting features in rack ensure proper heat flow during autoclaving and reduce condensation on tips
  • Hinged lid prevents sterile lid from coming in contact with lab bench
  • Finger notch on racks allows easy removal/insert of refill trays
  • Snap-and-tear bags make it easy to open each individually wrapped sterile rack
  • Well-labeled racks simplify record keeping, tip identification and re-ordering
  • Environmentally sensitive reduced rack size reduces waste
  • Racks utilize recycled material and are marked with appropriate symbols to support recycling initiatives
TRUSTED_SUSTAINABILITY

Especificaciones

Volume (Metric) 300 μL
Filtered Non-filtered
Sterility Non-sterile
Autoclavable Autoclavable
Format Bulk
Tip Style Extended Length
For Use With (Equipment) Universal Fit
Product Line Matrix TallTip
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado