missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scie Plas Maxi Standard Gel Electrophoresis Unit
For high-resolution analytical, preparative studies of nucleic acids
Brand: Scie Plas HU20
This item is not returnable.
View return policy
Description
- Large format
- Four comb slots along tray for faster separation
- Multi-channel pipette compatible combs
- Colour coded, height adjustable
- Buffer recirculation ports
- Coloured loading strips
- UV-transparent acrylic casting tray
- Side handles for safe, easy transportation around laboratory
- Unit dimensions (Lx W x H): 47.5 x 27 x 8cm
- No of combs: 2
- Comb thickness: 1, 1.5 or 2mm
- Comb throughput: 16 to 42 samples
- Comb slots: 4
- Migration distance between comb slots: 5cm
Specifications
Gel Electrophoresis Unit | |
1 x gel casting tray, 2 x 1.0mm 16 sample combs, power supply connectors and loading strips | |
168 | |
2200 mL |
220 x 200 mm | |
Maxi Standard | |
2200 mL | |
220 mm |