Learn More
Thermo Scientific™ Maxima H Minus Double-Stranded cDNA Synthesis Kit
The Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA from total RNA or mRNA.
Brand: Thermo Scientific™ K2563
4142.57 GBP valid until 2024-12-31
Use promo code "22010" to get your promotional price.
Description
Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA from total RNA or mRNA. First- and second-strand cDNA synthesis reactions are performed in the same tube without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains premixed components to reduce the number of pipetting steps necessary to complete the procedure.
First- and second-strand cDNA synthesis reactions are performed in the same tube without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains pre-mixed components to reduce the number of pipetting steps necessary to complete the procedure.
Features of the Maxima H Minus Double-Stranded cDNA Synthesis Kit include:
- Efficient synthesis of full-length double-stranded cDNA
- Fast—procedure completed in less than two hours
- Convenient—pre-mixed components
- Complete—includes all primers, controls and residual RNA removal reagents
Applications
- Full-length double-stranded blunt-end cDNA synthesis for cloning
- Double-stranded cDNA library construction
Specifications
-25°C to -15°C | |
50°C to 55°C | |
cDNA Synthesis Kit | |
Maxima H Minus | |
Double Stranded cDNA | |
Separate Components |
200 reactions | |
Maxima | |
200 Reactions | |
Cloning | |
200 Reactions | |
RNA |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.