missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Maxima H Minus Reverse Transcriptase Promotion
A highly thermostable, high processivity reverse transcriptase lacking RNase H activity
£46.26 - £153.00
Specifications
Content And Storage | -20°C |
---|---|
For Use With (Application) | cDNA Synthesis, RT-PCR, Gene Expression |
Concentration | 5X |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15767496
|
Thermo Scientific™
EP0751 |
2000 U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15777496
|
Thermo Scientific™
EP0752 |
10,000 U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Thermostable - 90% active after incubation at 50°C for 60 minutes in a reaction mixture.
- Active up to 65°C
- High yields of full-length cDNA up to 20 kb
- High sensitivity - reproducible cDNA synthesis from a wide range of starting total RNA amounts (1 pg to 5 μg)
- Efficient - complete cDNA synthesis in 15 to 30 minutes
- Increased resistance to common reaction inhibitors
- Incorporates modified nucleotides
Specifications
-20°C | |
cDNA Synthesis, RT-PCR, Gene Expression | |
5X |