missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Maximum Recovery Diluent

Product Code. p-7113339
Click to view available options
Fill Weight:
500.0g + 1.5g
9.0g + 0.3g
90.0g + 1.5g
Unit Size:
Pack of 10
Pack of 24
This item is not returnable. View return policy

Product Code. 12756949

Brand: Thermo Scientific™ BO0348D

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Achieve maximum recovery of microorganisms during dilution with this protective, isotonic diluent.

Achieve maximum recovery of microorganisms during dilution with this protective, isotonic Thermo Scientific™ Oxoid™ Maximum Recovery Diluent.

The presence of peptone in the diluent provides viable counts to ensure accurate quantitative procedures are performed. The isotonic strength of the diluent ensures recovery of organisms from various sources which may be vulnerable in distilled water or aqueous suspensions. The low peptone concentration in the diluent reduces organism multiplication in the sample for at least 45 minutes during the dilution stage1,2

Maintain the viability of organisms during dilution process with Maximum Recovery Diluent with low levels of peptone.

  • Allows accurate quantitative procedures to be performed with minimal reduction in viable count.
  • Reduces organism multiplication in the sample for at least one hour during the dilution stage.
  • Ready-to-use.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless, clear
Content And Storage 2 to 25°C
Description Maximum Recovery Diluent
Format Universal bottles
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 9.0g + 0.3g
Packaging Type Twenty four universal bottles with screw cap closures in a box
Quantity 24/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.