missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maximum Recovery Diluent

Product Code. 12703596
Click to view available options
Quantity:
100/Pk.
50/Pk.
Format:
Tube
Universal Tube
Fill Volume:
9 mL
Manufacturing Origin:
Germany
Scotland
Unit Size:
Pack of 100
Pack of 50
This item is not returnable. View return policy

Product Code. 12703596

Brand: Thermo Scientific™ EB0348D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximize your microorganism recovery with a protective and isotonic diluent in convenient ready-prepared tubes.

Maintain viability and maximize the recovery of microorganisms with protective and isotonic Thermo Scientific™ Maximum Recovery Diluent . Maximum Recovery Diluent combines the protective effect of peptone in the diluting solution1 with the osmotic support of physiological saline1,2.

The isotonic strength of Maximum Recovery Diluent ensures recovery of organisms from various sources which may be vulnerable in distilled water or aqueous suspensions.

The low concentration of peptone does not cause multiplication of the organisms within 1 to 2 hours of dilution of the sample.

Technique:

  1. Put 10g of the test sample into a sterile blender jar or sterile plastic bag.
  2. Add 90mL of sterile Maximum Recovery Diluent.
  3. Operate the blender according to its speed for sufficient time to give a total number of 15,000 to 20,000 revolutions. Alternatively, operate a peristaltic type blender (Stomacher) for 2 minutes.
  4. Within 15 minutes, transfer 1mL of the macerate to 9mL of sterile diluent and mix well (10-1 dilution).
  5. Prepare additional decimal dilutions in the same way.
  6. Aseptically transfer 1mL of each dilution of the initial suspension in duplicate to the center of a dish.
  7. Prepare pour plates with the medium of choice.
  8. Allow the agar to solidify and incubate.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Light ivory, transparent
Content And Storage 2°C to 10°C
Description Maximum Recovery Diluent
Format Universal Tube
Manufacturing Origin Scotland
Fill Volume 9 mL
Fill Weight 10 ± 0.5mL
Packaging Type 100 universals in a carton box
Quantity 100/Pk.

For In Vitro Diagnostic Use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.