Learn More
Fisherbrand™ MaxiThermal Software
MaxiThermal Software for USB Data Loggers.
Brand:  Fisherbrand™ 61010-96
Product Code. 15348764
  • £238.00 / Each
Estimated Shipment: 20-06-2025
to see stock.
Explore more special offers
Add to basket
Requires : Standard USB 1.1 interface
Description
Description
MaxiThermal Software for USB Data Loggers.
  • Graphic representations are a simple X – Y plot of time versus temperature with unlimited zoom, view temperature data points on the graph, view total elapsed time, or the exact time when it was recorded
  • Graph and all data points (date, time and temperature) can be printed, as well as the ability to add a user note to the file for including information pertinent to that particular plot
  • Ability to view numeric data in a screen window and Data can be Exported as a CSV file for use in other software
  • Summary Information report showing the specific logger information, the Start and Stop Time, Duration of Recording, Measurement Interval, Highest, Lowest, Median and Average Temperatures as well as the High and Low Alarm information
  • Data from the logger is easily downloaded to your PC via a standard USB 1.1 interface
  • Compatible with Windows 98, NT, 2000, ME, XP, Vista, and Windows 7 and 8
Specifications
Specifications
Datalogger Software
MaxiThermal
Viewing and transferring recorded data from loggers to computer
No Display
MaxiThermal Software CD
Fisherbrand Traceable Datalogging Logger-Trac Thermometers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ MaxiThermal Software > MaxiThermal Software