Learn More
Thermo Scientific™ Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Mix and match these shaker platform clamps, mats and racks, available in a variety of sizes, to suit individual needs in the laboratory.
Brand:  Thermo Scientific™ 75004104
 View more versions of this product
Product Code. 17871875
  • £587.38 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Universal Clamps, available in a variety of sizes, to suit individual needs in the laboratory. Compatible with MaxQ shaker platforms.
Mix and match Thermo Scientific™ Solaris™ shaker Universal clamps to suit individual needs in the laboratory.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vessel Clamp
11 x 14 in. (369 x 289 mm)
Single height adjustable clamp
Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers