Learn More
Thermo Scientific™ Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Mix and match these shaker platform clamps, mats and racks, available in a variety of sizes, to suit individual needs in the laboratory.
Brand:  Thermo Scientific™ 75004102
 View more versions of this product
Product Code. 17881875
  • £689.09 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Universal Clamps, available in a variety of sizes, to suit individual needs in the laboratory. Compatible with MaxQ shaker platforms.
Mix and match Thermo Scientific™ Solaris™ shaker Universal clamps to suit individual needs in the laboratory.  
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vessel Clamp
12 x 14 in.
Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers
387 x 326 mm
Dual Height adjstable clamp