Learn More
Thermo Scientific™ Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Mix and match these shaker platform clamps, mats and racks, available in a variety of sizes, to suit individual needs in the laboratory.
Brand:  Thermo Scientific™ 75004107
 View more versions of this product
Product Code. 17891875
  • £176.30 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Universal Clamps, available in a variety of sizes, to suit individual needs in the laboratory. Compatible with MaxQ shaker platforms.
Mix and match Thermo Scientific™ Solaris™ shaker Universal clamps to suit individual needs in the laboratory.  Adhesive mats are compatible with all Thermo Scientific™ Solaris and MaxQ platforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Media Bottle Clamp
87 x 87 x 98 mm
250 mL square media bottle clamp
Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers