Learn More
Thermo Scientific™ MaxQ™ Shaker Universal Platforms
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms.
Brand: Thermo Scientific™ 3522-3
105.34 GBP valid until 2024-12-31
Use promo code "21970" to get your promotional price.
This item is not returnable.
View return policy
Description
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.
For use with MaxQ 30100 platforms (11 × 13in.) and MaxQ 30110 platforms (18 × 18in.).Specifications
Stacking Adapter | |
13 x 11 in. (33 x 28 cm) | |
279.4 mm (MaxQ 30100 Platforms), 457.2 mm (MaxQ 30110 Platforms) | |
MaxQ™ 30100 and MaxQ™ 30110 Platform |
Certificate of Analysis (COA), Certificate of Conformance (COC) | |
11 in. (MaxQ 30100 Platforms), 18 in. (MaxQ 30110 Platforms) | |
Adapter set for stacking two platforms |