missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals May-Gruenwald Stain, pure

£31.60 - £93.30

Specifications

Chemical Name or Material May-Gruenwald Stain
CAS 524698-43-9
MDL Number MFCD00131580
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10216970
View Documents
Thermo Scientific Chemicals
194830250
25 g Glass bottle
£31.60
25g
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
10494755
View Documents
Thermo Scientific Chemicals
194831000
100 g Glass bottle
£93.30
100g
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

May-Gruenwald Stain
MFCD00131580
Authentic
Pure
Brown or Green
524698-43-9
Solubility in water: soluble.
in 66:33 IMS:water, 655 to 670nm (blue), 515 to 530nm (red)
Glass bottle
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • May-Gruenwald Stain

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only