missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ MB22 Premier Disposable Low-profile Microtome Blade
Choose the Epredia™ MB22 premier disposable low-profile microtome blade for applications where extremely thin, high-quality sections are required.
Brand: Epredia™ 3050822
170.52 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Low-profile
- Increased durability
Specifications
Low Profile Blade | |
Designed with a steeper angle that allows users to cut thinner sections | |
22° | |
0.25 mm |
Low | |
Single-use | |
0.01 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction