Learn More
Gibco™ MBP Bovine Protein, Natural
MBP Bovine Protein, Natural
Brand: Gibco™ 13228010
Code : 50
Additional Details : Weight : 0.10000kg
Description
Myelin Basic Protein (MBP) was purified from bovine brain by a modification of the method of Deibler et al. (1). The protein is supplied lyophilized.Specifications
10 mg | |
Store in freezer (-5 to -30°C). | |
Powder |
Bovine | |
Approved for shipment on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.