Learn More
Thermo Scientific™ MBTFA Acylation Reagent
Shop All Thermo Scientific ProductsDescription
Trifluoroacylate primary and secondary amines, hydroxyl and thiol groups, and carbohydrates with the Thermo Scientific™ MBTFA Acylation Reagent. The principal byproduct N-methyltrifluoroacetamine is stable, volatile, and does not present problems in subsequent GC. MBTFA produces very volatile derivatives of carbohydrates and can be used to selectively acylate amines in the presence of hydroxyl and carboxyl groups that have been protected by silylation.
- Reacts under nonacidic conditions
- Principle byproduct from the derivatization reaction is N-methyltrifluoroacetamine, which is stable, volatile and does not present problems in subsequent GC
- Produces very volatile derivatives of carbohydrates
- Can be used to selectively acylate amines in the presence of hydroxyl and carboxyl groups that have been protected by silylation
Specifications
Specifications
| For Use With | GC |
| Hazards | Corrosive, Flammable |
| Storage | -20°C |
| Quantity | 5 g |
| Standard Components | N-Methyl-bis(trifluoroacetamide) |
| Standard Concentration | 98 to 100% |
| Item Description | 5 g |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.