missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MBTFA Acylation Reagent

Trifluoroacylate primary and secondary amines, hydroxyl and thiol groups, and carbohydrates with the Thermo Scientific™ MBTFA Acylation Reagent.

Brand:  Thermo Scientific™ TS-49700


 View more versions of this product

Product Code. 11757057

  • £269.34 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Trifluoroacylate primary and secondary amines, hydroxyl and thiol groups, and carbohydrates with the Thermo Scientific™ MBTFA Acylation Reagent. The principal byproduct N-methyltrifluoroacetamine is stable, volatile, and does not present problems in subsequent GC. MBTFA produces very volatile derivatives of carbohydrates and can be used to selectively acylate amines in the presence of hydroxyl and carboxyl groups that have been protected by silylation.

  • Reacts under nonacidic conditions
  • Principle byproduct from the derivatization reaction is N-methyltrifluoroacetamine, which is stable, volatile and does not present problems in subsequent GC
  • Produces very volatile derivatives of carbohydrates
  • Can be used to selectively acylate amines in the presence of hydroxyl and carboxyl groups that have been protected by silylation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GC
-20°C
N-Methyl-bis(trifluoroacetamide)
10 x 1 mL ampules
Corrosive, Flammable
10 x 1 mL ampules
98 to 100%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use to extinguish.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.