missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBTPS2, Mouse anti-Human, Clone: 1A3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00051360-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: ASTLQQLSFPVRAYKRLDGSTECCNNHSLTDVCFSYRNNFNKRLHTCLPARKAVEATQVCRTNKDCKKSSSSSFCIIPSLETHTRLIKVKHPPQIDMLYVGHPLHLHSpecifications
MBTPS2 | |
Monoclonal | |
Unconjugated | |
membrane-bound transcription factor peptidase, site 2 | |
FLJ32174/S2P | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Western Blot | |
1A3 | |
In 1x PBS, pH 7.4 | |
NM_015884 | |
MBTPS2 | |
MBTPS2 (NP_056968.1, 312 a.a. ∼ 418 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
51360 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction