missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MC3R Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21027-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a G-protein-coupled receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone that is expressed in tissues other than the adrenal cortex and melanocytes. This gene maps to the same region as the locus for benign neonatal epilepsy. Mice deficient for this gene have increased fat mass despite decreased food intake, suggesting a role for this gene product in the regulation of energy homeostasis. Mutations in this gene are associated with a susceptibility to obesity in humans.Specifications
MC3R | |
Polyclonal | |
Unconjugated | |
MC3R | |
BMIQ9, MC3, MC3 R, MC3R, melanocortin 3 receptor, Melanocortin receptor 3, OB20, OQTL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4159 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P41968 | |
MC3R | |
MC3R Fusion Protein Ag15292 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |