missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ McCoy 5A Medium, Iwakata and Grace Modification with L-Glutamine

Ideal for leukocytes, biopsy tissues, a broad range of human and rat normal and transformed cell types, and demanding primary and continuous cell lines

Brand:  MP Biomedicals™ 091255354

Product Code. 17378593

  • £34.30 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

McCoy's media were originally formulated for the growth and support of human lymphocytes. The 5A modification originated as a reformulation of McCoy's by Hsu and Kellogg. It was then further modified by Iwakata and Grace featuring increased levels of i-inositol and glucose. This final modification produced a media identical to RPMI 1629. McCoy's 5A media supports the indefinite proliferation of Walker 256 carcinoma cells.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 4°C. Protect from light.
Liquid
Sterile
Mammalian Cell Culture
500 mL
McCoy 5A Medium
Cell Culture Media
No Glutamine
6.6 to 7.2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.