missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ McCoy's 5A Medium (Park and Terasaki modification)
GREENER_CHOICE

Product Code. 11574526
Click to view available options
Quantity:
1 Ea.
Unit Size:
100mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11574526

Brand: Gibco™ 80014020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues.

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.

This McCoy's 5A (modified) is provided as follows:.

With: High Glucose, L-glutamine, Bacto-peptone, Phenol Red.

Without: Sodium Pyruvate.

The complete formulation is available.

Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. Park and Terasaki modified McCoy's medium with modified levels of glycine, L-glutamic acid, L-hydroxyproline, L-proline, L-serine, L-threonine, L-tryptophan, and L-valine, plus the addition of antibiotics and HEPES.

Product Use.

For Research Use Only. Not for use in diagnostic procedures.

Gibco™ McCoy's 5A (Park and Terasaki modification) requires serum supplementation, commonly with 10% Fetal Bovine Serum (FBS). Gibco™ McCoy's 5A (Park and Terasaki modification) uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Liquid
Product Type McCoy's 5A Medium
Sterility Sterile-filtered
With Additives High Glucose, Glutamine, HEPES, Phenol Red
Without Additives No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Quantity 1 Ea.
Content And Storage Storage conditions:2°C to 8°C. Protect from light.
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.