missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ McCoy's 5A (Modified) Medium, HEPES
GREENER_CHOICE

Product Code. p-7076080
Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
500mL
Pack of 10
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10674413

Brand: Gibco™ 22330021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

McCoy's 5A (Modified) Medium, HEPES

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.

Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. This product also includes Dr. Hsu's addition of Hanks' salts to enable use outside a CO2 incubator.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ McCoy's 5A is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ McCoy's 5A product made in our Grand Island facility (12330-031). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Gibco™ McCoy's 5A (modified) requires serum supplementation, commonly with 10% Fetal Bovine Serum (FBS). Gibco™ McCoy's 5A (modified) uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Liquid
Product Type McCoy's 5A (Modified) Medium
With Additives High Glucose, Glutamine, Bacto-peptone, Phenol Red, HEPES
Without Additives No Sodium Pyruvate
For Use With (Application) Mammalian cell culture
Quantity 500 mL
Certifications/Compliance ISO 13485 standards, FDA
pH 7 to 7.4
Regulatory Status For in-vitro diagnostic use
Content And Storage 2 to 8°C
Description McCoy's 5A (Modified) Medium, 500mL, 1 X Concentration, Liquid Form, With High Glucose, L-glutamine, Bacto-peptone, Phenol Red, HEPES, 7 to 7.4 pH, 12 Months Shelf Life, 285 to 325mOsm/kg Osmolality, Used for Mammalian Cell Culture
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.