missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MCDB 131 Medium, no glutamine
GREENER_CHOICE

Product Code. 11513407
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11513407

Brand: Gibco™ 10372019

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

MCDB 131 Medium, no glutamine

MCDB 131 Medium was originally developed by Knedler and Ham as reduced serum-supplemented medium for the culture of human microvascular endothelial cells (HUVEC). MCDB 131 Medium is also used with other cell types, including human omental microvascular cells, hepatocytes, myocytes, and smooth muscle cells.

Using MCDB 131
MCDB 131 Medium contains many components not found in traditional basal media, such as trace elements, putrescine, adenine, thymidine, and higher levels of some amino acids and vitamins. These additions allow the medium to be supplemented with very low levels of serum or defined components. MCDB 131 Medium contains no proteins or growth factors, and is often supplemented with EGF, hydrocortisone, glutamine, and low levels of Fetal Bovine Serum (FBS). The FBS concentration must be optimized for each cell type. MCDB 131 Medium uses a sodium bicarbonate buffer system (1.176 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Dual-site cGMP manufacturing
For supply chain continuity, we manufacture MCDB 131 at two separate facilities, located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP-manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Human Microvascular Endothelial Cells (HMVEC), Human Omental Microvascular Cells, Hepatocytes, Myocytes, Smooth Muscle Cells
Concentration 1 X
Form Liquid
Product Type MCDB 131 Medium
Sterility Sterile-filtered
With Additives Low Glucose, Phenol Red, Sodium Pyruvate
Without Additives No Glutamine, No HEPES
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2–8°C (protect from light)
Shelf life: 12 months from date of manufacture

Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.