missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Antibody, Novus Biologicals™
Chicken Polyclonal Antibody has been used in 26 publications
£179.00 - £494.00
Specifications
| Antigen | mCherry |
|---|---|
| Dilution | Western Blot 1:2000-1:5000, ELISA, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, IHC |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Free Floating), Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18069453
|
Novus Biologicals
NBP2-25158-0.025ML |
0.025 mL |
£179.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18736713
|
Novus Biologicals
NBP2-25158 |
£494.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
mCherry Polyclonal specifically detects mCherry in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount.Specifications
| mCherry | |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Free Floating), Immunofluorescence, Western Blot | |
| Unconjugated | |
| Chicken | |
| DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:2000-1:5000, ELISA, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, IHC | |
| Polyclonal | |
| Precipitation | |
| RUO | |
| IgY | |
| Ammonium sulfate precipitation | |
| This antibody detects mCherry. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title