missing translation for 'onlineSavingsMsg'
Learn More

mCherry Mouse, Clone: 1B3C6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 68088-1-IG-150UL

Product Code. 17619776

This item is not returnable. View return policy

Description

Description

mCherry Monoclonal antibody apecifically detetects mCherry antigen in Recombinant Protein Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

mCherry
Monoclonal
1000 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
mCherry fusion protein Ag25320
150 μL
Store at -20°C. Stable for one year after shipment.
IgG1
Western Blot, ELISA
1B3C6
Unconjugated
Mouse
Protein G purification
Primary
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers